2-Chloro-5-vinylpyrimidine ≥98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with TBC

A766416-100MG A766416-250MG
77901-491EA 40 USD
77901-491 77902-038
2-Chloro-5-vinylpyrimidine ≥98% stabilized
2-Chloro-5-vinylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂
MDL Number: MFCD11040635
CAS Number: 131467-06-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR