4-Bromo-2-chloronicotinic acid ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A766138-5G A766138-1G A766138-25G
77684-704EA 166 USD
77684-704 77893-514 77893-515
4-Bromo-2-chloronicotinic acid ≥97%, Ambeed.Inc
4-Bromo-2-chloronicotinic acid

Precaución: For research use only.

Formula: C₆H₃BrClNO₂
MDL Number: MFCD13189211
CAS Number: 1060805-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR