2-Iodo-5-methylpyrazine ≥95%

Supplier: Ambeed
Warning

A766102-250MG A766102-100MG A766102-5G
77674-104EA 32 USD
77674-104 77882-660 77882-661
2-Iodo-5-methylpyrazine ≥95%
2-Iodo-5-methylpyrazine

Precaución: for Research Use Only

Formula: C₅H₅IN₂
MDL Number: MFCD08705755
CAS Number: 914452-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR