1-(((tert-Butoxycarbonyl)amino)methyl)cyclopropane-1-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A765385-1G A765385-250MG A765385-5G
77884-695EA 82 USD
77884-695 77884-696 77884-697
1-(((tert-Butoxycarbonyl)amino)methyl)cyclopropane-1-carboxylic acid ≥97%
1-(((tert-Butoxycarbonyl)amino)methyl)cyclopropane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₄
MDL Number: MFCD04973108
CAS Number: 204376-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR