(5aS,10bR)-2-mesityl-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium chloride hydrate ≥97%
Supplier: Ambeed
|
Warning
|
A765267-100MG
A765267-1G
A765267-250MG
77622-910EA
120.51
USD
77622-910
77622-912
77622-914
(5aS,10bR)-2-mesityl-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium chloride hydrate ≥97%
(5aS,10bR)-2-mesityl-4,5a,6,10b-tetrahydroindeno[2,1-b][1,2,4]triazolo[4,3-d][1,4]oxazin-2-ium chloride hydrate
Precaución: for Research Use Only
Formula:
C₂₁H₂₄ClN₃O₂ MW: 385.89 g/mol |
MDL Number:
MFCD20265586 CAS Number: 2002470-81-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...