4-(tert-Butyl)pyridine 1-oxide ≥98%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butyl)pyridine N-oxide, 4-(2-Methyl-2-propanyl)pyridine 1-oxide, 4-tert-Butyl-1-oxypyridine, 4-tert-Butylpyridine 1-oxide

A764794-250MG A764794-1G A764794-5G A764794-100MG
77783-992EA 59 USD
77783-992 77783-991 77783-993 77783-990
4-(tert-Butyl)pyridine 1-oxide ≥98%
4-tert-Butylpyridine-1-oxide

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00082431
CAS Number: 23569-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR