4-Chloro-2-(dimethylamino)nicotinaldehyde ≥98%

Supplier: Ambeed
Warning

A764411-1G A764411-5G
77882-656EA 19 USD
77882-656 77882-657
4-Chloro-2-(dimethylamino)nicotinaldehyde ≥98%
4-Chloro-2-(dimethylamino)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O
MW: 184.63 g/mol
MDL Number: MFCD12025925
CAS Number: 1160474-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR