3-Cyano-2-methoxy-5-pyridineboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

A762949-1G A762949-5G
77873-210EA 154 USD
77873-210 77873-211
3-Cyano-2-methoxy-5-pyridineboronic acid pinacol ester ≥95%
3-Cyano-2-methoxy-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₃
MDL Number: MFCD11504968
CAS Number: 1073354-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR