2-(2-(2-(2-(Aminooxy)ethoxy)ethoxy)ethoxy)ethanol ≥95%

Supplier: Ambeed
Danger

Synonyms: Aminooxy-PEG4-alcohol

A761994-250MG A761994-5G
77622-892EA 59.74 USD
77622-892 77783-983
2-(2-(2-(2-(Aminooxy)ethoxy)ethoxy)ethoxy)ethanol ≥95%
2-(2-(2-(2-(Aminooxy)ethoxy)ethoxy)ethoxy)ethanol

Precaución: for Research Use Only

Formula: C₈H₁₉NO₅
MW: 209.24 g/mol
MDL Number: MFCD30828691
CAS Number: 106492-60-8
UN: 1760
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR