Biotin-HPDP ≥95%

Supplier: Ambeed
Warning

A761726-100MG A761726-250MG A761726-50MG
77622-888EA 312.09 USD
77622-888 77622-890 77783-982
Biotin-HPDP ≥95%
Biotin-HPDP

Precaución: for Research Use Only

Formula: C₂₄H₃₇N₅O₃S₃
MDL Number: MFCD00078541
CAS Number: 129179-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR