Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate) ≥95%

Supplier: Ambeed
Warning

A761354-5G A761354-25G
77622-880EA 256.47 USD
77622-880 77783-981
Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate) ≥95%
Tris(1,10-phenanthroline)cobalt(III) tris(hexafluorophosphate)

Precaución: for Research Use Only

Formula: C₃₆H₂₄CoF₁₈N₆P₃
MW: 1034.46 g/mol
MDL Number: MFCD30749311
CAS Number: 28277-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR