4-(4-(5,5-Dimethyl-4,5-dihydrothiazol-2-yl)piperazin-1-yl)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine ≥95%

Supplier: Ambeed
Warning

A761249-1MG A761249-10MG A761249-5MG A761249-100MG A761249-50MG
77614-350EA 47.38 USD
77614-350 77686-769 77686-770 77893-511 77900-877
4-(4-(5,5-Dimethyl-4,5-dihydrothiazol-2-yl)piperazin-1-yl)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine ≥95%
4-(4-(5,5-Dimethyl-4,5-dihydrothiazol-2-yl)piperazin-1-yl)-6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₇H₂₀F₃N₅S₂
MDL Number: MFCD29044864
CAS Number: 1454920-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR