1-Butyl-1-methylpyrrolidinium bis(fluorosulfonyl)imide ≥98%
Supplier: Ambeed
|
Warning
|
A761058-1G
A761058-5G
77622-872EA
12.36
USD
77622-872
77622-874
1-Butyl-1-methylpyrrolidinium bis(fluorosulfonyl)imide ≥98%
1-Butyl-1-methylpyrrolidinium bis(fluorosulfonyl)imide
Precaución: for Research Use Only
Formula:
C₉H₂₀F₂N₂O₄S₂ MW: 322.39 g/mol |
MDL Number:
MFCD30749250 CAS Number: 1057745-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...