2,5-Dioxopyrrolidin-1-yl 4,7,10,13,16,19,22,25-octaoxaoctacos-27-yn-1-oate ≥95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG8-NHS ester

A760335-250MG A760335-100MG A760335-1G
77599-018EA 63.55 USD
77599-018 77622-856 77622-858
2,5-Dioxopyrrolidin-1-yl 4,7,10,13,16,19,22,25-octaoxaoctacos-27-yn-1-oate ≥95%
2,5-Dioxopyrrolidin-1-yl 4,7,10,13,16,19,22,25-octaoxaoctacos-27-yn-1-oate

Precaución: for Research Use Only

Formula: C₂₄H₃₉NO₁₂
MW: 533.5660 g/mol
MDL Number: MFCD31536764
CAS Number: 2182601-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR