2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulfonate ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-N-PEG2-Ms

A760076-5G A760076-1G A760076-250MG
77595-202EA 228.58 USD
77595-202 77622-852 77622-854
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulfonate ≥95%
2-(2-((tert-Butoxycarbonyl)amino)ethoxy)ethyl methanesulfonate

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₆S
MW: 283.3418 g/mol
MDL Number: MFCD24465573
CAS Number: 302331-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR