3-(Methylamino)propanal, N-BOC protected ≥96%
Supplier: Ambeed
|
Warning
|
A760064-250MG
A760064-100MG
77674-086EA
110.35
USD
77674-086
77882-652
3-(Methylamino)propanal, N-BOC protected ≥96%
3-(Methylamino)propanal, N-BOC protected
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₃ |
MDL Number:
MFCD09751791 CAS Number: 273757-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...