3-(Cyanomethyl)benzeneboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetonitrile, 2-[3-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile

A759941-1G A759941-25G A759941-5G
77670-052EA 13 USD
77670-052 77873-205 77873-206
3-(Cyanomethyl)benzeneboronic acid pinacol ester ≥98%
3-(Cyanomethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.113 g/mol
MDL Number: MFCD02179458
CAS Number: 396131-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR