1-tert-Butyl 5-(N-Succinimidyl) N-Palmitoyl-L-glutamate ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Palmitoyl-L-glutamic acid 1-tert-Butyl 5-(N-Succinimidyl) ester, Pal-Glu(OSu)-OtBu, PAlm-Glu(NHS)-OtBu

A759910-1G A759910-5G
77599-016EA 39.98 USD
77599-016 77622-850
1-tert-Butyl 5-(N-Succinimidyl) N-Palmitoyl-L-glutamate ≥98%
1-tert-Butyl 5-(N-Succinimidyl) N-Palmitoyl-L-glutamate

Precaución: for Research Use Only

Formula: C₂₉H₅₀N₂O₇
MDL Number: MFCD22989465
CAS Number: 204521-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR