2-(Isobutyrylamino)acetic acid ≥95%

Supplier: Ambeed
Warning

A759849-5G A759849-1G
77674-082EA 197 USD
77674-082 77882-651
2-(Isobutyrylamino)acetic acid ≥95%
2-(Isobutyrylamino)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₃
MDL Number: MFCD09043666
CAS Number: 15926-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR