5-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%

Supplier: Ambeed
Warning

A759601-1G A759601-100MG
77627-512EA 42.25 USD
77627-512 77878-600
5-Chloro-2-hydroxy-3-methylbenzaldehyde ≥97%
5-Chloro-2-hydroxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD00275687
CAS Number: 23602-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR