tert-Butyl 4-amino-3-methoxybenzoate 95+%

Supplier: Ambeed
Warning

A759234-1G A759234-5G
77670-050EA 699 USD
77670-050 77873-203
tert-Butyl 4-amino-3-methoxybenzoate 95+%
tert-Butyl 4-amino-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MDL Number: MFCD08703410
CAS Number: 123330-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR