(1S,2S,3S,5S)-3-(Benzyloxy)-5-(6-(benzyloxy)-2-(((4-methoxyphenyl)diphenylmethyl)amino)-9H-purin-9-yl)-2-((benzyloxy)methyl)cyclopentanol ≥95%

Supplier: Ambeed
Warning

A759218-1G A759218-5G
77674-078EA 31 USD
77674-078 77674-080
(1S,2S,3S,5S)-3-(Benzyloxy)-5-(6-(benzyloxy)-2-(((4-methoxyphenyl)diphenylmethyl)amino)-9H-purin-9-yl)-2-((benzyloxy)methyl)cyclopentanol ≥95%
(1S,2S,3S,5S)-3-(Benzyloxy)-5-(6-(benzyloxy)-2-(((4-methoxyphenyl)diphenylmethyl)amino)-9H-purin-9-yl)-2-((benzyloxy)methyl)cyclopentanol

Precaución: for Research Use Only

Formula: C₅₂H₄₉N₅O₅
MW: 823.97 g/mol
MDL Number: MFCD11978033
CAS Number: 142217-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR