NVS-SM1 ≥98%

Supplier: Ambeed
Warning

Synonyms: (6Z)-3-(1H-pyrazol-4-yl)-6-[3-(2,2,6,6-tetramethylpiperidin-4-yl)oxy-1H-pyridazin-6-ylidene]cyclohexa-2,4-dien-1-one, 5-(1H-Pyrazol-4-yl)-2-(6-((2,2,6,6-tetramethylpiperidin-4-yl)oxy)pyridazin-3-yl)phenol, Branaplam, LMI070

A758913-250MG A758913-50MG A758913-100MG A758913-1G
77747-496EA 583 USD
77747-496 77747-497 77747-494 77747-495
NVS-SM1 ≥98%
NVS-SM1

Precaución: for Research Use Only

Formula: C₂₂H₂₇N₅O₂
MW: 393.49 g/mol
MDL Number: MFCD29472288
CAS Number: 1562338-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR