(1R,3S)-3-Aminocyclohexanol hydrochloride ≥98%

Supplier: Ambeed
Warning

A758845-250MG A758845-5G
77573-448EA 62 USD
77573-448 77893-507
(1R,3S)-3-Aminocyclohexanol hydrochloride ≥98%
(1R,3S)-3-Aminocyclohexanol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MDL Number: MFCD22689399
CAS Number: 1849594-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR