3-Bromo-1-(phenylsulfonyl)indole ≥97%

Supplier: Ambeed
Warning

A758222-5G A758222-1G
77614-344EA 40.24 USD
77614-344 77893-501
3-Bromo-1-(phenylsulfonyl)indole ≥97%
3-Bromo-1-(phenylsulfonyl)indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO₂S
MDL Number: MFCD02681986
CAS Number: 99655-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR