Benzyl 4-(2-(tert-butoxycarbonyl)hydrazinyl)piperidine-1-carboxylate ≥95%

Supplier: Ambeed
Warning

A757905-10G A757905-1G
77686-768EA 248 USD
77686-768 77893-500
Benzyl 4-(2-(tert-butoxycarbonyl)hydrazinyl)piperidine-1-carboxylate ≥95%
Benzyl 4-(2-(tert-butoxycarbonyl)hydrazinyl)piperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₇N₃O₄
MW: 349.424 g/mol
MDL Number: MFCD12407285
CAS Number: 280111-50-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR