(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid ≥98%

Supplier: Ambeed
Warning

A757682-1G A757682-250MG
77573-440EA 306 USD
77573-440 77882-648
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid ≥98%
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₂₈N₄O₄
MDL Number: MFCD02684483
CAS Number: 823780-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR