nitro-Grela ≥97%

Supplier: Ambeed
Warning

A757504-250MG A757504-100MG
77618-094EA 127.72 USD
77618-094 77747-490
nitro-Grela ≥97%
nitro-Grela

Precaución: for Research Use Only

Formula: C₃₁H₃₇Cl₂N₃O₃Ru
MDL Number: MFCD28411645
CAS Number: 502964-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR