Methyltetrazine-nhs ester ≥95%

Supplier: Ambeed
Warning

A757270-100MG A757270-1G A757270-250MG A757270-50MG A757270-5G
77599-004EA 148.62 USD
77599-004 77599-006 77599-008 77622-842 77622-844
Methyltetrazine-nhs ester ≥95%
Methyltetrazine-nhs ester

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₅O₄
MDL Number: MFCD28334557
CAS Number: 1644644-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR