2-(tert-Butoxycarbonylamino)pentanoic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-Nva-OH, 2-((tert-Butoxycarbonyl)amino)pentanoic acid

A757124-25G A757124-5G
77783-957EA 41 USD
77783-957 77783-958
2-(tert-Butoxycarbonylamino)pentanoic acid ≥95%
2-(tert-Butoxycarbonylamino)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD00270725
CAS Number: 521286-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR