VX-702 ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Difluorophenyl)-6-(1-(2,6-difluorophenyl)ureido)nicotinamide, 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide

A756758-1MG A756758-100MG A756758-1G A756758-250MG
77614-340EA 6.18 USD
77614-340 77686-764 77686-765 77686-766
VX-702 ≥99%
VX-702

Precaución: for Research Use Only

Formula: C₁₉H₁₂F₄N₄O₂
MW: 404.32 g/mol
MDL Number: MFCD11616590
CAS Number: 745833-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR