2-Methyl-1-((2-methyl-5-nitrophenyl)sulfonyl)-1H-benzo[d]imidazole ≥97%

Supplier: Ambeed
Warning

Synonyms: BI-6015

A756299-50MG A756299-100MG
77747-486EA 64 USD
77747-486 77747-485
2-Methyl-1-((2-methyl-5-nitrophenyl)sulfonyl)-1H-benzo[d]imidazole ≥97%
2-Methyl-1-((2-methyl-5-nitrophenyl)sulfonyl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃O₄S
MW: 331.34 g/mol
MDL Number: MFCD04406654
CAS Number: 93987-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR