DBCO-C6-NHS ester ≥95%

Supplier: Ambeed
Warning

Synonyms: DBCO-NHS ester 2, Dbco-c6-nhs ester

A756218-100MG A756218-250MG A756218-1G
77598-988EA 115.83 USD
77598-988 77598-990 77622-816
DBCO-C6-NHS ester ≥95%
DBCO-C6-NHS ester

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₂O₅
MW: 430.453 g/mol
MDL Number: MFCD22380763
CAS Number: 1384870-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR