3-(Methanesulfonylmethyl)aniline ≥98%

Supplier: Ambeed
Warning

A756129-250MG A756129-1G
77783-943EA 45 USD
77783-943 77783-942
3-(Methanesulfonylmethyl)aniline ≥98%
3-(Methanesulfonylmethyl)aniline

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂S
MDL Number: MFCD12776627
CAS Number: 261925-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR