Mal-PEG3-NHS ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dioxopyrrolidin-1-yl 3-(2-(2-(2-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethoxy)ethoxy)ethoxy)propanoate

A756028-1G A756028-250MG A756028-100MG
77622-814EA 380.21 USD
77622-814 77783-941 77783-940
Mal-PEG3-NHS ester ≥95%
Mal-PEG3-NHS ester

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₉
MDL Number: MFCD26127782
CAS Number: 1537892-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR