2-(2-Bromoethoxy)acetic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: Bromo-PEG1-CH2COOH

A756022-100MG A756022-250MG A756022-5G
77622-808EA 42.23 USD
77622-808 77622-810 77622-812
2-(2-Bromoethoxy)acetic acid ≥98%
2-(2-Bromoethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₄H₇BrO₃
MW: 183.0006 g/mol
MDL Number: MFCD20637665
CAS Number: 1135131-50-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR