Cbz-N-amido-PEG3-acid ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-NH-PEG3-C2-acid, CBz-n-amido-peg3-acid
A755988-1G
A755988-100MG
A755988-250MG
77573-438EA
174
USD
77573-438
77622-804
77622-806
Cbz-N-amido-PEG3-acid ≥95%
Cbz-N-amido-PEG3-acid
Precaución: for Research Use Only
Formula:
C₁₇H₂₅NO₇ |
MDL Number:
MFCD28142484 CAS Number: 1310327-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...