2-Fluoro-3-(trifluoromethyl)benzoyl chloride ≥98%

Supplier: Ambeed
Danger

A755801-1G A755801-5G
77901-566EA 29 USD
77901-566 77901-156
2-Fluoro-3-(trifluoromethyl)benzoyl chloride ≥98%
2-Fluoro-3-(trifluoromethyl)benzoyl chloride

Precaución: for Research Use Only

Formula: C₈H₃ClF₄O
MDL Number: MFCD00061154
CAS Number: 208173-19-7
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR