Thiol-PEG8-acid ≥98%

Supplier: Ambeed
Warning

A755751-250MG A755751-100MG
77783-936EA 201 USD
77783-936 77783-935
Thiol-PEG8-acid ≥98%
Thiol-PEG8-acid

Precaución: for Research Use Only

Formula: C₁₉H₃₈O₁₀S
MDL Number: MFCD08705300
CAS Number: 866889-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR