tert-Butyl 1-bromo-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate ≥95%

Supplier: Ambeed
Warning

Synonyms: Bromo-PEG8-Boc

A755436-100MG A755436-250MG A755436-5G
77598-982EA 62.53 USD
77598-982 77622-800 77622-802
tert-Butyl 1-bromo-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate ≥95%
tert-Butyl 1-bromo-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oate

Precaución: for Research Use Only

Formula: C₂₃H₄₅BrO₁₀
MW: 561.5 g/mol
MDL Number: MFCD26793748
CAS Number: 1623792-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR