tert-Butyl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate ≥95%

Supplier: Ambeed
Warning

Synonyms: Propargyl-PEG2-Boc

A755344-5G A755344-100MG A755344-1G A755344-250MG
77622-796EA 611.82 USD
77622-796 77622-790 77622-792 77622-794
tert-Butyl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate ≥95%
tert-Butyl 3-(2-(prop-2-yn-1-yloxy)ethoxy)propanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MDL Number: MFCD28505565
CAS Number: 1807503-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR