O-(2-(2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)hydroxylamine ≥95%

Supplier: Ambeed
Danger

Synonyms: Aminooxy-PEG3-propargyl

A755105-100MG A755105-1G A755105-250MG A755105-5G
77598-966EA 53.3 USD
77598-966 77598-968 77598-970 77622-780
O-(2-(2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)hydroxylamine ≥95%
O-(2-(2-(2-(Prop-2-yn-1-yloxy)ethoxy)ethoxy)ethyl)hydroxylamine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MDL Number: MFCD28950771
CAS Number: 1807537-27-4
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR