3-Oxo-1-phenyl-2,7,10,13,16,19,22-heptaoxa-4-azapentacosan-25-oic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: Cbz-NH-PEG6-C2-acid

A755064-250MG A755064-1G
77598-964EA 44.08 USD
77598-964 77622-778
3-Oxo-1-phenyl-2,7,10,13,16,19,22-heptaoxa-4-azapentacosan-25-oic acid ≥95%
3-Oxo-1-phenyl-2,7,10,13,16,19,22-heptaoxa-4-azapentacosan-25-oic acid

Precaución: for Research Use Only

Formula: C₂₃H₃₇NO₁₀
MW: 487.54 g/mol
MDL Number: MFCD13184966
CAS Number: 1334177-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR