Fmoc-N-amido-PEG8-NHS ester ≥95%

Supplier: Ambeed
Warning

A755054-1G A755054-5G
77783-925EA 150 USD
77783-925 77783-926
Fmoc-N-amido-PEG8-NHS ester ≥95%
Fmoc-N-amido-PEG8-NHS ester

Precaución: for Research Use Only

Formula: C₃₈H₅₂N₂O₁₄
MDL Number: MFCD21363318
CAS Number: 1334170-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR