Biotin-PEG6-alcohol ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(17-Hydroxy-3,6,9,12,15-pentaoxaheptadecyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

A755037-250MG A755037-50MG A755037-100MG
77783-923EA 217 USD
77783-923 77783-924 77783-922
Biotin-PEG6-alcohol ≥98%
Biotin-PEG6-alcohol

Precaución: for Research Use Only

Formula: C₂₂H₄₁N₃O₈S
MDL Number: MFCD26142990
CAS Number: 906099-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR