28-Bromo-2,5,8,11,14,17,20,23,26-nonaoxaoctacosane ≥95%

Supplier: Ambeed
Warning

Synonyms: m-PEG9-Br

A754529-1G A754529-5G
77598-944EA 205 USD
77598-944 77622-766
28-Bromo-2,5,8,11,14,17,20,23,26-nonaoxaoctacosane ≥95%
28-Bromo-2,5,8,11,14,17,20,23,26-nonaoxaoctacosane

Precaución: for Research Use Only

Formula: C₁₉H₃₉BrO₉
MDL Number: MFCD28950767
CAS Number: 125562-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR