Mal-PEG5-acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3,6,9,12,15-pentaoxaoctadecan-18-oic acid

A754471-100MG A754471-50MG
77622-762EA 182.31 USD
77622-762 77622-764
Mal-PEG5-acid ≥98%
Mal-PEG5-acid

Precaución: for Research Use Only

Formula: C₁₇H₂₇NO₉
MW: 389.398 g/mol
MDL Number: MFCD25424108
CAS Number: 1286755-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR