5-Methyl-1,3,4-oxadiazole-2-thiol ≥97%
Supplier: Ambeed
|
Warning
|
A754406-1G
A754406-100MG
77627-500EA
65.92
USD
77627-500
77878-595
5-Methyl-1,3,4-oxadiazole-2-thiol ≥97%
5-Methyl-1,3,4-oxadiazole-2-thiol
Precaución: for Research Use Only
Formula:
C₃H₄N₂OS |
MDL Number:
MFCD03161246 CAS Number: 31130-17-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...