N-(2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide ≥95%

Supplier: Ambeed
Warning

A754212-250MG A754212-100MG
77783-909EA 167 USD
77783-909 77783-908
N-(2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide ≥95%
N-(2-(2-(2-(2-Hydroxyethoxy)ethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide

Precaución: for Research Use Only

Formula: C₁₈H₃₃N₃O₆S
MDL Number: MFCD03701139
CAS Number: 1778736-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR