TBDMS-PEG4-Acid ≥95%

Supplier: Ambeed
Danger

Synonyms: 2,2,3,3-Tetramethyl-4,7,10,13-tetraoxa-3-silahexadecan-16-oic acid

A754096-100MG A754096-1G A754096-250MG
77622-746EA 58.71 USD
77622-746 77622-748 77622-750
TBDMS-PEG4-Acid ≥95%
TBDMS-PEG4-Acid

Precaución: for Research Use Only

Formula: C₁₅H₃₂O₆Si
MW: 336.496 g/mol
MDL Number: MFCD26793772
CAS Number: 1807540-77-7
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR